Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142809.1 | NP_001136281.1 | 3 - 4 | 1300 | 78 | |
NM_001142810.1 | NP_001136282.1 | 3 - 4 | 1300 | 78 | ||
NM_133987.2 | NP_598748.1 | 3 - 4 | 1300 | 78 | ||
GenBank mRNA | AB077327.1 | - | 3 - 4 | 648 | 78 | |
AF459435.1 | - | 3 - 4 | 1283 | 78 | ||
BC024444.1 | - | 2 - 3 | 313 | 78 | ||
BC049801.1 | - | 3 - 4 | 475 | 78 | ||
BC141067.1 | - | 4 - 5 | 1041 | 9055679 | 78 | |
BC145319.1 | - | 4 - 5 | 705 | 9053874 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |