Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029868.2 | NP_001025039.1 | 2 - 3 | 936 | 68 | |
GenBank mRNA | BC056462.1 | - | 2 - 3 | 282 | 5703229 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |