Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007823.2 | NP_031849.1 | 5 - 6 | 641 | 80 | |
GenBank mRNA | AK078882.1 | - | 2 - 3 | 253 | 80 | |
AK143890.1 | - | 5 - 6 | 654 | 80 | ||
AK165445.1 | - | 5 - 6 | 657 | 80 | ||
BC008996.1 | - | 5 - 6 | 645 | 3967680 | 80 | |
D50834.1 | - | 5 - 6 | 641 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |