Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007823.2 | NP_031849.1 | 7 - 8 | 899 | 69 | |
GenBank mRNA | AK078882.1 | - | 4 - 5 | 511 | 69 | |
AK143890.1 | - | 7 - 8 | 912 | 69 | ||
AK165445.1 | - | 7 - 8 | 915 | 69 | ||
BC008996.1 | - | 7 - 8 | 903 | 3967680 | 69 | |
D50834.1 | - | 7 - 8 | 899 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |