Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005511.3 | NP_001005511.2 | 7 - 8 | 717 | 62 | |
NM_001290990.1 | NP_001277919.1 | 6 - 7 | 872 | 62 | ||
NR_110987.1 | 7 - 8 | 717 | 62 | |||
GenBank mRNA | AB288873.1 | - | 7 - 8 | 711 | 62 | |
BC057032.1 | - | 6 - 7 | 605 | 62 | ||
BC059845.1 | - | 7 - 8 | 926 | 62 | ||
BC083185.1 | - | 6 - 7 | 872 | 6855048 | 62 | |
BC094377.1 | - | 7 - 8 | 717 | 30536822 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |