Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC138049.1 | - | 20 - 21 | 2761 | 8861068 | 87 |
BC138050.1 | - | 20 - 21 | 2761 | 8861069 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |