Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_001164292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_024192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
NM_024192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014251.mm.1 |
TC1900001600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164295.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_001164295.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_024192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
NM_024192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014249.mm.1 |
TC1900001600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164291.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164291.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164295.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_001164295.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_024192.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
NM_024192.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900014248.mm.1 |
TC1900001600.mm.1 |
MTA TC |