Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081067.1 | NP_001074536.1 | 2 - 3 | 85 | 62 | |
GenBank mRNA | BC080745.1 | - | 420 | 62 | ||
DQ520794.1 | - | 2 - 3 | 81 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |