Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500012137.mm.1 |
NM_028992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012137.mm.1 |
NM_028992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012137.mm.1 |
TC1500002345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012140.mm.1 |
NM_028992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012140.mm.1 |
NM_028992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012140.mm.1 |
TC1500002345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012138.mm.1 |
NM_028992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012138.mm.1 |
NM_028992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012138.mm.1 |
TC1500002345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012141.mm.1 |
NM_028992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012141.mm.1 |
NM_028992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012141.mm.1 |
TC1500002345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012139.mm.1 |
NM_028992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012139.mm.1 |
NM_028992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012139.mm.1 |
TC1500002345.mm.1 |
MTA TC |