Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC104048.1 | - | 3 - 4 | 386 | 40040143 | 82 |
BC104049.2 | - | 3 - 4 | 386 | 40040144 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |