Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133791.4 | NP_598552.2 | 2 - 3 | 593 | 65 | |
GenBank mRNA | AB045323.1 | - | 2 - 3 | 616 | 65 | |
AK079375.1 | - | 2 - 3 | 428 | 65 | ||
AK086956.1 | - | 2 - 3 | 421 | 65 | ||
AK147648.1 | - | 587 | 65 | |||
AK147944.1 | - | 2 - 3 | 562 | 65 | ||
BC067050.1 | - | 2 - 3 | 593 | 30537299 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |