Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1cf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081074.1 | NP_001074543.1 | 8 - 9 | 1286 | 66 | |
GenBank mRNA | AY566862.1 | - | 8 - 9 | 1283 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |