Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146180.1 | NP_001139652.1 | 9 - 10 | 1326 | 74 | |
NM_144800.2 | NP_659049.2 | 10 - 11 | 1338 | 74 | ||
GenBank mRNA | AK030533.1 | - | 8 - 9 | 732 | 74 | |
AK129139.1 | - | 9 - 10 | 1323 | 74 | ||
AY214918.1 | - | 10 - 11 | 839 | 74 | ||
BC023682.1 | - | 4 - 5 | 444 | 74 | ||
BC024131.1 | - | 10 - 11 | 1303 | 5101888 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |