Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033481.1 | NP_001028653.1 | 14 - 15 | 2163 | 120 | |
GenBank mRNA | AK039649.1 | - | 4 - 5 | 447 | 120 | |
AK141695.1 | - | 14 - 15 | 2164 | 120 | ||
BC157942.1 | - | 11 - 12 | 1554 | 9007277 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |