Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF229633.1 | - | 11 - 12 | 946 | 99 | |
AK122481.2 | - | 9 - 10 | 805 | 99 | ||
AK134989.1 | - | 11 - 12 | 1580 | 99 | ||
BC086631.1 | - | 12 - 13 | 1490 | 99 | ||
U61363.1 | - | 3 - 4 | 349 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |