Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF229633.1 | - | 13 - 14 | 1278 | 57 | |
AK122481.2 | - | 11 - 12 | 1137 | 57 | ||
AK134989.1 | - | 13 - 14 | 1912 | 57 | ||
BC086631.1 | - | 14 - 15 | 1822 | 57 | ||
BU149633.1 | - | 316 | 57 | |||
U61363.1 | - | 5 - 6 | 681 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |