Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028195.3 | NP_082471.2 | 3 - 4 | 74 | ||
GenBank mRNA | AF079972.1 | - | 3 - 4 | 254 | 74 | |
AK036679.1 | - | 3 - 4 | 74 | |||
AK152306.1 | - | 3 - 4 | 74 | |||
AK152909.1 | - | 3 - 4 | 74 | |||
AK157026.1 | - | 3 - 4 | 229 | 74 | ||
AK220198.1 | - | 3 - 4 | 252 | 74 | ||
BC139054.1 | - | 3 - 4 | 296 | 8862076 | 74 | |
BC139056.1 | - | 3 - 4 | 296 | 8862078 | 74 | |
BC145528.1 | - | 3 - 4 | 296 | 74 | ||
BY161747.1 | - | 3 - 4 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |