Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800016015.mm.1 |
NM_001081150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016015.mm.1 |
NM_001081150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016015.mm.1 |
TC0800002060.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016014.mm.1 |
NM_001081150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016014.mm.1 |
NM_001081150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016014.mm.1 |
TC0800002060.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800016016.mm.1 |
NM_001081150.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016016.mm.1 |
NM_001081150.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800016016.mm.1 |
TC0800002060.mm.1 |
MTA TC |