Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030292.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030293.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030291.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030298.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030294.mm.1 |
NM_010365.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
TC0500003369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
NM_001080746.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
NM_001080747.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
NM_001080748.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
NM_001080749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030295.mm.1 |
NM_010365.4 |
Refseq |