Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmtk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081109.1 | NP_001074578.1 | 4 - 5 | 688 | 57 | |
GenBank mRNA | AB288872.1 | - | 4 - 5 | 516 | 57 | |
AK158724.1 | - | 4 - 5 | 689 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mus musculus Probe spans exons |