Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081066.1 | NP_001074535.1 | 8 - 9 | 1386 | 115 | |
GenBank mRNA | AK173046.1 | - | 8 - 9 | 1286 | 115 | |
BC132389.1 | - | 8 - 9 | 1274 | 40130666 | 115 | |
BC137791.1 | - | 8 - 9 | 1274 | 8860810 | 115 | |
BC144912.1 | - | 8 - 9 | 1274 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |