Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500004630.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004630.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004630.mm.1 |
TC1500000640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004632.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004632.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004632.mm.1 |
TC1500000640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004633.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004633.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004633.mm.1 |
TC1500000640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004629.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004629.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004629.mm.1 |
TC1500000640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004631.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004631.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004631.mm.1 |
TC1500000640.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500004628.mm.1 |
NM_008677.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004628.mm.1 |
NM_008677.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500004628.mm.1 |
TC1500000640.mm.1 |
MTA TC |