Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011537.3 | NP_035667.1 | 7 - 8 | 1402 | 74 | |
GenBank mRNA | AF140427.1 | - | 7 - 8 | 1167 | 74 | |
AK044843.1 | - | 3 - 4 | 381 | 74 | ||
BC090639.1 | - | 7 - 8 | 1406 | 30548234 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |