Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164724.1 | NP_001158196.1 | 4 - 5 | 411 | 62 | |
NM_133775.2 | NP_598536.2 | 4 - 5 | 390 | 62 | ||
GenBank mRNA | AK020353.1 | - | 4 - 5 | 391 | 62 | |
AK075849.1 | - | 4 - 5 | 412 | 62 | ||
AK163464.1 | - | 4 - 5 | 391 | 62 | ||
AY905582.1 | - | 3 - 4 | 331 | 62 | ||
BC003847.1 | - | 4 - 5 | 367 | 3593927 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |