Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081308.2 | NP_001074777.1 | 9 - 10 | 1017 | 88 | |
NM_001199685.1 | NP_001186614.1 | 10 - 11 | 1125 | 88 | ||
NM_183306.2 | NP_899129.2 | 10 - 11 | 1102 | 88 | ||
GenBank mRNA | AK040531.1 | - | 10 - 11 | 1126 | 88 | |
AK042181.1 | - | 10 - 11 | 1025 | 88 | ||
AK155208.1 | - | 9 - 10 | 1013 | 88 | ||
AK171353.1 | - | 9 - 10 | 3123 | 88 | ||
BC158072.1 | - | 10 - 11 | 1067 | 9088167 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |