Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081308.2 | NP_001074777.1 | 13 - 14 | 1612 | 62 | |
NM_001199685.1 | NP_001186614.1 | 14 - 15 | 1720 | 62 | ||
NM_183306.2 | NP_899129.2 | 14 - 15 | 1697 | 62 | ||
GenBank mRNA | AK039665.1 | - | 1 - 2 | 270 | 62 | |
AK042181.1 | - | 14 - 15 | 1620 | 62 | ||
AK155208.1 | - | 13 - 14 | 1608 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mus musculus Probe spans exons |