Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081308.2 | NP_001074777.1 | 5 - 6 | 626 | 78 | |
NM_001199685.1 | NP_001186614.1 | 6 - 7 | 734 | 78 | ||
NM_183306.2 | NP_899129.2 | 6 - 7 | 711 | 78 | ||
GenBank mRNA | AK040531.1 | - | 6 - 7 | 735 | 78 | |
AK042181.1 | - | 6 - 7 | 634 | 78 | ||
AK155208.1 | - | 5 - 6 | 621 | 78 | ||
BC158072.1 | - | 6 - 7 | 676 | 9088167 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |