Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164639.1 | NP_001158111.1 | 4 - 5 | 688 | 106 | |
NM_009289.3 | NP_033315.2 | 4 - 5 | 688 | 106 | ||
GenBank mRNA | AF039574.1 | - | 4 - 5 | 693 | 106 | |
AF112855.1 | - | 4 - 5 | 577 | 106 | ||
AK029491.1 | - | 4 - 5 | 555 | 106 | ||
AK122218.1 | - | 4 - 5 | 1029 | 106 | ||
BC131674.1 | - | 4 - 5 | 589 | 40110915 | 106 | |
BC131675.1 | - | 4 - 5 | 589 | 40110921 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |