Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159650.1 | NP_001153122.1 | 4 - 5 | 388 | 59 | |
NM_021352.3 | NP_067327.1 | 4 - 5 | 458 | 59 | ||
GenBank mRNA | AJ272229.2 | - | 3 - 4 | 442 | 59 | |
AK053552.1 | - | 2 - 3 | 772 | 59 | ||
BC031442.1 | - | 3 - 4 | 449 | 4501690 | 59 | |
BC058522.1 | - | 3 - 4 | 367 | 6824229 | 59 | |
CJ046471.1 | - | 4 - 5 | 459 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |