Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013774.2 | NP_001013796.2 | 2 - 3 | 208 | 69 | |
GenBank mRNA | AY950703.1 | - | 2 - 3 | 209 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |