Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500032890.mm.1 |
TC0500003587.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032890.mm.1 |
NM_001013774.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032890.mm.1 |
NM_001013774.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032889.mm.1 |
TC0500003587.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032889.mm.1 |
NM_001013774.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032889.mm.1 |
NM_001013774.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032884.mm.1 |
TC0500003587.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032884.mm.1 |
NM_001013774.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032884.mm.1 |
NM_001013774.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032888.mm.1 |
TC0500003587.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032888.mm.1 |
NM_001013774.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032888.mm.1 |
NM_001013774.2 |
Refseq |