Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133738.1 | NP_598499.1 | 6 - 7 | 972 | 95 | |
GenBank mRNA | AK155460.1 | - | 6 - 7 | 1055 | 95 | |
AK166124.1 | - | 744 | 95 | |||
AK170655.1 | - | 6 - 7 | 610 | 95 | ||
AK171292.1 | - | 6 - 7 | 647 | 95 | ||
BC076595.1 | - | 6 - 7 | 972 | 30649897 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |