Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ide | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031156.3 | NP_112419.3 | 13 - 14 | 1767 | 81 | |
GenBank mRNA | AJ278422.1 | - | 13 - 14 | 1787 | 81 | |
AK128986.1 | - | 13 - 14 | 1784 | 81 | ||
BC041675.1 | - | 13 - 14 | 1799 | 4951971 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |