Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020044.3 | NP_064428.1 | 9 - 10 | 566 | 94 | |
NM_022964.4 | NP_075253.2 | 8 - 9 | 530 | 94 | ||
GenBank mRNA | AF257136.1 | - | 8 - 9 | 510 | 94 | |
AK134721.1 | - | 6 - 7 | 258 | 94 | ||
AK138953.1 | - | 9 - 10 | 525 | 94 | ||
AK143533.1 | - | 8 - 9 | 531 | 94 | ||
AK144261.1 | - | 9 - 10 | 505 | 94 | ||
AK162321.1 | - | 8 - 9 | 489 | 94 | ||
AY190024.1 | - | 9 - 10 | 531 | 94 | ||
BC005804.1 | - | 8 - 9 | 459 | 3154073 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |