Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcn1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172719.2 | NP_766307.2 | 28 - 29 | 3335 | 52 | |
GenBank mRNA | AB093217.1 | - | 4 - 5 | 528 | 52 | |
AK089324.1 | - | 2 - 3 | 196 | 52 | ||
AK090137.1 | - | 28 - 29 | 3333 | 52 | ||
AK147257.1 | - | 28 - 29 | 3328 | 52 | ||
BC082548.1 | - | 15 - 16 | 2113 | 52 | ||
BC150735.1 | - | 28 - 29 | 3338 | 9087646 | 52 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |