Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080798.1 | NP_001074267.1 | 3 - 4 | 1152 | 69 | |
NM_133919.3 | NP_598680.3 | 3 - 4 | 1163 | 69 | ||
GenBank mRNA | AF074266.1 | - | 3 - 4 | 996 | 69 | |
AK082878.1 | - | 4 - 5 | 1220 | 69 | ||
AK143742.1 | - | 3 - 4 | 1152 | 69 | ||
AY749168.1 | - | 3 - 4 | 996 | 69 | ||
BC131665.1 | - | 3 - 4 | 996 | 40110795 | 69 | |
CD543840.1 | - | 1 - 2 | 277 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |