Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033561.1 | NP_291039.1 | 3 - 4 | 340 | 68 | |
GenBank mRNA | AK045004.1 | - | 3 - 4 | 342 | 68 | |
AK088818.1 | - | 3 - 4 | 352 | 68 | ||
AK122194.1 | - | 3 - 4 | 340 | 68 | ||
AK132034.1 | - | 3 - 4 | 360 | 68 | ||
AK135390.1 | - | 3 - 4 | 341 | 68 | ||
AK136999.1 | - | 3 - 4 | 327 | 68 | ||
AK149841.1 | - | 3 - 4 | 326 | 68 | ||
AK150088.1 | - | 3 - 4 | 349 | 68 | ||
AK166567.1 | - | 3 - 4 | 341 | 68 | ||
AK168872.1 | - | 3 - 4 | 359 | 68 | ||
AK169312.1 | - | 3 - 4 | 323 | 68 | ||
AK169353.1 | - | 3 - 4 | 341 | 68 | ||
AK169407.1 | - | 3 - 4 | 359 | 68 | ||
AK177188.1 | - | 2 - 3 | 101 | 68 | ||
BC014796.1 | - | 3 - 4 | 354 | 3962104 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |