Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009879.3 | NP_034009.2 | 3 - 4 | 353 | 88 | |
GenBank mRNA | AF354757.1 | - | 3 - 4 | 353 | 88 | |
AK013424.1 | - | 3 - 4 | 347 | 88 | ||
AK161375.1 | - | 3 - 4 | 342 | 88 | ||
BC055721.1 | - | 3 - 4 | 310 | 6810406 | 88 | |
Y10495.2 | - | 3 - 4 | 353 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |