Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038837.1 | - | 2758 | 61 | ||
BC132355.1 | - | 2822 | 40130632 | 61 | ||
BC132357.1 | - | 2822 | 40130634 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus |