Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164573.1 | NP_001158045.1 | 20 - 21 | 2094 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |