Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172276.3 | NP_758480.2 | 11 - 12 | 1866 | 76 | |
GenBank mRNA | AK031872.1 | - | 11 - 12 | 1857 | 76 | |
AK088182.1 | - | 11 - 12 | 1810 | 76 | ||
AK140369.1 | - | 11 - 12 | 1859 | 76 | ||
BC023268.1 | - | 6 - 7 | 897 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |