Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec31b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033343.1 | NP_001028515.1 | 8 - 9 | 974 | 72 | |
GenBank mRNA | AK158017.1 | - | 7 - 8 | 851 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |