Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025391.2 | NP_001020562.1 | 10 - 11 | 1498 | 137 | |
NM_015752.3 | NP_056567.2 | 10 - 11 | 1501 | 137 | ||
GenBank mRNA | AF134893.1 | - | 10 - 11 | 1441 | 137 | |
AJ131692.1 | - | 10 - 11 | 1499 | 137 | ||
AJ308626.1 | - | 10 - 11 | 1432 | 137 | ||
AK016976.1 | - | 10 - 11 | 1479 | 137 | ||
AK156410.1 | - | 10 - 11 | 1463 | 137 | ||
BC048168.1 | - | 10 - 11 | 1471 | 6821477 | 137 | |
BC056997.1 | - | 10 - 11 | 1471 | 6821477 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |