Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028544.1 | NP_082820.1 | 5 - 6 | 1871 | 82 | |
GenBank mRNA | AK033276.1 | - | 3 - 4 | 1159 | 82 | |
AK156602.1 | - | 5 - 6 | 1883 | 82 | ||
BC072584.1 | - | 2 - 3 | 825 | 82 | ||
BC127059.1 | - | 3 - 4 | 797 | 40109852 | 82 | |
BC150766.1 | - | 5 - 6 | 1871 | 9087677 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |