Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028544.1 | NP_082820.1 | 7 - 8 | 2061 | 85 | |
GenBank mRNA | AK011557.2 | - | 1 - 2 | 144 | 85 | |
AK033276.1 | - | 5 - 6 | 1349 | 85 | ||
AK156602.1 | - | 7 - 8 | 2073 | 85 | ||
BC028444.1 | - | 133 | 85 | |||
BC072584.1 | - | 4 - 5 | 1015 | 85 | ||
BC127059.1 | - | 5 - 6 | 987 | 40109852 | 85 | |
BC150766.1 | - | 7 - 8 | 2061 | 9087677 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |