Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163540.1 | NP_001157012.1 | 15 - 16 | 2350 | 123 | |
NM_001163542.1 | NP_001157014.1 | 16 - 17 | 1867 | 123 | ||
NM_001163549.1 | NP_001157021.1 | 15 - 16 | 1913 | 123 | ||
NM_001164203.1 | NP_001157675.1 | 16 - 17 | 1854 | 123 | ||
NM_011117.2 | NP_035247.2 | 14 - 15 | 1816 | 123 | ||
NM_201385.2 | NP_958787.2 | 14 - 15 | 1751 | 123 | ||
NM_201386.2 | NP_958788.2 | 14 - 15 | 1812 | 123 | ||
NM_201387.3 | NP_958789.3 | 14 - 15 | 1841 | 123 | ||
NM_201388.2 | NP_958790.2 | 14 - 15 | 1803 | 123 | ||
NM_201389.2 | NP_958791.2 | 14 - 15 | 2335 | 123 | ||
NM_201390.2 | NP_958792.2 | 14 - 15 | 1750 | 123 | ||
NM_201391.2 | NP_958793.2 | 14 - 15 | 1898 | 123 | ||
NM_201392.2 | NP_958794.2 | 14 - 15 | 1724 | 123 | ||
NM_201393.2 | NP_958795.2 | 14 - 15 | 1841 | 123 | ||
NM_201394.2 | NP_958796.2 | 14 - 15 | 1898 | 123 | ||
GenBank mRNA | AF188008.1 | - | 15 - 16 | 2350 | 123 | |
AF188011.1 | - | 15 - 16 | 1913 | 123 | ||
AF188012.1 | - | 14 - 15 | 1898 | 123 | ||
AF188013.1 | - | 16 - 17 | 1867 | 123 | ||
AF188014.1 | - | 14 - 15 | 1816 | 123 | ||
AF188021.1 | - | 14 - 15 | 1724 | 123 | ||
AF188023.1 | - | 16 - 17 | 1854 | 123 | ||
AK017743.1 | - | 14 - 15 | 1731 | 123 | ||
AK044559.1 | - | 1911 | 123 | |||
AY480033.1 | - | 14 - 15 | 1816 | 123 | ||
AY480034.1 | - | 14 - 15 | 1751 | 123 | ||
AY480035.1 | - | 14 - 15 | 1812 | 123 | ||
AY480036.1 | - | 14 - 15 | 1841 | 123 | ||
AY480037.1 | - | 14 - 15 | 1803 | 123 | ||
AY480038.1 | - | 14 - 15 | 2335 | 123 | ||
AY480039.1 | - | 14 - 15 | 1750 | 123 | ||
AY480040.1 | - | 14 - 15 | 1898 | 123 | ||
AY480041.1 | - | 14 - 15 | 1724 | 123 | ||
AY480042.1 | - | 14 - 15 | 1841 | 123 | ||
AY480043.1 | - | 14 - 15 | 1898 | 123 | ||
BC021810.1 | - | 14 - 15 | 1885 | 123 | ||
BC099929.1 | - | 14 - 15 | 2336 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |