Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168288.1 | NP_001161760.1 | 9 - 10 | 3135 | 75 | |
NM_198420.2 | NP_940812.2 | 8 - 9 | 3042 | 75 | ||
GenBank mRNA | AK035479.1 | - | 8 - 9 | 2840 | 75 | |
AK173232.1 | - | 8 - 9 | 2977 | 75 | ||
BC060065.1 | - | 4 - 5 | 1614 | 75 | ||
BC060637.1 | - | 8 - 9 | 3023 | 6849400 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |