Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022430.2 | NP_071875.2 | 4 - 5 | 522 | 72 | |
GenBank mRNA | AF237906.1 | - | 4 - 5 | 460 | 72 | |
AK008099.1 | - | 4 - 5 | 461 | 72 | ||
AY039038.1 | - | 4 - 5 | 460 | 72 | ||
BC042719.1 | - | 4 - 5 | 508 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |