Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028595.4 | NP_082871.2 | 3 - 4 | 480 | 152 | |
GenBank mRNA | AF237910.1 | - | 3 - 4 | 438 | 152 | |
AK150397.1 | - | 3 - 4 | 377 | 152 | ||
AK152258.1 | - | 3 - 4 | 377 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |