Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133768.4 | NP_598529.1 | 8 - 9 | 688 | 91 | |
GenBank mRNA | AK076453.1 | - | 8 - 9 | 703 | 91 | |
AK138423.1 | - | 8 - 9 | 685 | 91 | ||
AK140160.1 | - | 8 - 9 | 703 | 91 | ||
AK148764.1 | - | 8 - 9 | 672 | 91 | ||
AK168739.1 | - | 8 - 9 | 683 | 91 | ||
BC016670.1 | - | 8 - 9 | 688 | 2648753 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |